• USA Home
  • Search Results

  • ethylmaltol14014494011811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl maltol

4 Products
Synonym:   2-Ethyl-3-hydroxy-4H-pyran-4-one, Ethyl maltol
Image of Ethyl maltol
CAS Number:
Empirical Formula (Hill Notation):
 C7H8O3
Molecular Weight:
 140.14
 Council of Europe no.:
 692
 EC Number:
 FEMA Number:
 3487
 Flavis number:
 7.047
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Anal. Standard Form
  Physical Form
  Titertest
  Assay
  Special Grade
Show More