• USA Home
  • Search Results

  • ethylmethanesulfonate124166250011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl methanesulfonate

3 Products
Synonym:   Ethyl mesylate, Methanesulfonic acid ethyl ester
Image of Ethyl methanesulfonate
CAS Number:
Linear Formula:
 CH3SO3CH2CH3
Molecular Weight:
 124.16
 Beilstein/REAXYS Number:
 773969
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Application(s)
Show More