• USA Home
  • Search Results

  • ethylmyristate2564212406111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl myristate

4 Products
Synonym:   Ethyl tetradecanoate, Myristic acid ethyl ester
Image of Ethyl myristate
CAS Number:
Linear Formula:
 CH3(CH2)12COOC2H5
Molecular Weight:
 256.42
 Beilstein/REAXYS Number:
 1776382
 Council of Europe no.:
 385
 EC Number:
 FEMA Number:
 2445
 Flavis number:
 9.104
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Refractive Index
  Document Links
  Allergen
  Organoleptic property
  Agency/Method
  Special Grade
  Assay
Show More