• USA Home
  • Search Results

  • ethylnonanoate1862912329511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl nonanoate

5 Products
Synonym:   Ethyl pelargonate
Image of Ethyl nonanoate
CAS Number:
Linear Formula:
 CH3(CH2)7COOC2H5
Molecular Weight:
 186.29
 Beilstein/REAXYS Number:
 1759169
 Council of Europe no.:
 388
 EC Number:
 FEMA Number:
 2447
 Flavis number:
 9.107
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Organoleptic property
  Application(s)
  Agency/Method
  Solubility
  Special Grade
  Assay
Show More