• USA Home
  • Search Results

  • ethylpalmitate2844862897711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl palmitate

6 Products
Synonym:   Ethyl hexadecanoate, Palmitic acid ethyl ester
Image of Ethyl palmitate
CAS Number:
Linear Formula:
 CH3(CH2)14COOC2H5
Molecular Weight:
 284.48
 Beilstein/REAXYS Number:
 1782663
 Council of Europe no.:
 634
 EC Number:
 FEMA Number:
 2451
 Flavis number:
 9.193
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Titertest
  Assay
  Special Grade
Show More