• USA Home
  • Search Results

  • ethylparaben1661712047811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethylparaben

3 Products
Synonym:   Ethyl 4-hydroxybenzoate, Ethylparaben
Image of Ethylparaben
CAS Number:
Linear Formula:
 HOC6H4CO2C2H5
Molecular Weight:
 166.17
 Beilstein/REAXYS Number:
 1101972
 E Number:
 E214
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID:
 eCl@ss
 39024522
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
Show More