• USA Home
  • Search Results

  • ethylperfluorobutyrate2420935627411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl perfluorobutyrate

1 Products
Synonym:   Ethyl heptafluorobutyrate
Image of Ethyl perfluorobutyrate
CAS Number:
Linear Formula:
 CF3CF2CF2COOCH2CH3
Molecular Weight:
 242.09
 Beilstein/REAXYS Number:
 1794634
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More