• USA Home
  • Search Results

  • ethylphenylacetate1642010197311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl phenylacetate

4 Products
Image of Ethyl phenylacetate
CAS Number:
Linear Formula:
 C6H5CH2COOC2H5
Molecular Weight:
 164.20
 Beilstein/REAXYS Number:
 509140
 Council of Europe no.:
 2156
 EC Number:
 FEMA Number:
 2452
 Flavis number:
 9.784
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Application(s)
  Special Grade
  Agency/Method
  Assay
Show More