• USA Home
  • Search Results

  • ethylpiperazinoacetate172224000408811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl piperazinoacetate

1 Products
Synonym:   1-(Ethoxycarbonylmethyl)piperazine, N-(Carboethoxymethyl)-piperazine, Ethyl 1-piperazineacetate
Image of Ethyl piperazinoacetate
CAS Number:
Empirical Formula (Hill Notation):
 C8H16N2O2
Molecular Weight:
 172.22
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More