• USA Home
  • Search Results

  • ethyls4chloro3hydroxybutyrate166608672885011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl (S)-(−)-4-chloro-3-hydroxybutyrate

1 Products
Image of Ethyl (<I>S</I>)-(−)-4-chloro-3-hydroxybutyrate
CAS Number:
Linear Formula:
 ClCH2CH(OH)CH2CO2C2H5
Molecular Weight:
 166.60
 Beilstein/REAXYS Number:
 4657170
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More