• USA Home
  • Search Results

  • ethyltrans3hexenoate14220239683011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl trans-3-hexenoate

2 Products
Image of Ethyl <I>trans</I>-3-hexenoate
CAS Number:
Linear Formula:
 C2H5CH=CHCH2CO2C2H5
Molecular Weight:
 142.20
 EC Number:
 FEMA Number:
 3342
 Flavis number:
 9.191
 MDL number:
 PubChem Substance ID:
Popular Documents:  Bulletin (PDF)  |  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More