• USA Home
  • Search Results

  • ethyltriphenylphosphoniumbromide37125153032111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyltriphenylphosphonium bromide

1 Products
Synonym:   TEP
Image of Ethyltriphenylphosphonium bromide
CAS Number:
Linear Formula:
 [(C6H5)3P+C2H5]Br-
Molecular Weight:
 371.25
 Beilstein/REAXYS Number:
 3599630
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More