• USA Home
  • Search Results

  • ethyltriphenylphosphoniumiodide41825473660111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyltriphenylphosphonium iodide

1 Products
Synonym:   ETPPI, Phenylphosphonium ethyl iodide
Image of Ethyltriphenylphosphonium iodide
CAS Number:
Linear Formula:
 (C6H5)3P(I)C2H5
Molecular Weight:
 418.25
 Beilstein/REAXYS Number:
 3659323
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More