• USA Home
  • Search Results

  • ethylvanillin1661712132411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl vanillin

4 Products
Synonym:   3-Ethoxy-4-hydroxybenzaldehyde, Ethylvanillin, NSC 1803, NSC 67240
Image of Ethyl vanillin
CAS Number:
Linear Formula:
 C2H5OC6H3(OH)CHO
Molecular Weight:
 166.17
 Beilstein/REAXYS Number:
 1073761
 Council of Europe no.:
 108
 EC Number:
 FEMA Number:
 2464
 Flavis number:
 5.019
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Assay
  Physical Form
  Agency/Method
  Application(s)
Show More