• USA Home
  • Search Results

  • ethylvinylether721110992211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethyl vinyl ether

3 Products
Synonym:   Ethoxyethylene
Image of Ethyl vinyl ether
CAS Number:
Linear Formula:
 C2H5OCH=CH2
Molecular Weight:
 72.11
 Beilstein/REAXYS Number:
 605351
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
 eCl@ss:
 39021025
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Potency
  Additive
Show More