• USA Home
  • Search Results

  • ethynyltrimethylsilane9822106654211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ethynyltrimethylsilane

1 Products
Synonym:   Trimethylsilylacetylene
Image of Ethynyltrimethylsilane
CAS Number:
Linear Formula:
 (CH3)3SiC≡CH
Molecular Weight:
 98.22
 Beilstein/REAXYS Number:
 906752
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More