• USA Home
  • Search Results

  • etodolac287354134025411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Etodolac

4 Products
Synonym:   1,8-Diethyl-1,3,4,9-tetrahydropyrano[3,4-b]indole-1-acetic acid, Etodolic acid, NSC 282126
Image of Etodolac
CAS Number:
Empirical Formula (Hill Notation):
 C17H21NO3
Molecular Weight:
 287.35
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More