• USA Home
  • Search Results

  • etoposide588563341942011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Etoposide

4 Products
Synonym:   4′-Demethylepipodophyllotoxin 9-(4,6-O-ethylidene-β-D-glucopyranoside), VP-16-213
Image of Etoposide
CAS Number:
Empirical Formula (Hill Notation):
 C29H32O13
Molecular Weight:
 588.56
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Mode of Action
  Physical Form
  Special Grade
Show More