• USA Home
  • Search Results

  • eugenol164209753011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Eugenol

11 Products
Synonym:   2-Methoxy-4-(2-propenyl)phenol, 4-Allyl-2-methoxyphenol, 4-Allylguaiacol
Image of Eugenol
CAS Number:
Linear Formula:
 4-(H2C=CHCH2)C6H3-2-(OCH3)OH
Molecular Weight:
 164.20
 Beilstein/REAXYS Number:
 1366759
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Shelf Life
  Anal. Standard Form
  Product Line
  Manufacturer designation
  Application(s)
  Assay
  Agency/Method
  Physical Form
Show More