• USA Home
  • Search Results

  • exendin441865714175874911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Exendin-4

1 Products
CAS Number:
Empirical Formula (Hill Notation):
 C184H282N50O60S
Molecular Weight:
 4186.57
 MDL number:
 NACRES:
 NA.32
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
E7144 Exendin-4, ≥97%,