• USA Home
  • Search Results

  • ezmixtryptone123459107940211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

EZMix Tryptone

1 Products
Synonym:   Peptone from casein
CAS Number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More