• USA Home
  • Search Results

  • famphur325345285711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Famphur

2 Products
Synonym:   O-(4-Dimethylsulfamoylphenyl)-O,O-dimethyl-phosphorothioate, Famophos
Image of Famphur
CAS Number:
Empirical Formula (Hill Notation):
 C10H16NO5PS2
Molecular Weight:
 325.34
 Beilstein/REAXYS Number:
 2224254
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Shelf Life
  Special Grade
Show More