• USA Home
  • Search Results

  • farnesol22237460284011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Farnesol

4 Products
Synonym:   3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol
Image of Farnesol
CAS Number:
Linear Formula:
 (CH3)2C=CHCH2CH2C(CH3)=CHCH2CH2C(CH3)=CHCH2OH
Molecular Weight:
 222.37
 Beilstein/REAXYS Number:
 1763926
 Council of Europe no.:
 78
 EC Number:
 FEMA Number:
 2478
 Flavis number:
 2.029
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Shelf Life
  Additive
  Titertest
  Special Grade
Show More