• USA Home
  • Search Results

  • fatbrownb29233653542811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fat Brown B

1 Products
Synonym:   Solvent red 3
Image of Fat Brown B
CAS Number:
Empirical Formula (Hill Notation):
 C18H16N2O2
Molecular Weight:
 292.33
 Colour Index Number:
 12010
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
F3257 Fat Brown B,