• USA Home
  • Search Results

  • fenbufen254283633085511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fenbufen

3 Products
Synonym:   γ-Oxo-(1,1′-biphenyl)-4-butanoic acid
Image of Fenbufen
CAS Number:
Linear Formula:
 C6H5C6H4CO(CH2)2CO2H
Molecular Weight:
 254.28
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
  Manufacturer designation
Show More