• USA Home
  • Search Results

  • fenbutatinoxide1052681335608611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fenbutatin oxide

1 Products
Synonym:   Bis[tris-(2-methyl-2-phenylpropyl)tin] oxide
Image of Fenbutatin oxide
CAS Number:
Empirical Formula (Hill Notation):
 C60H78OSn2
Molecular Weight:
 1052.68
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More