• USA Home
  • Search Results

  • fenpropimorph303486756491411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fenpropimorph

1 Products
Image of Fenpropimorph
CAS Number:
Empirical Formula (Hill Notation):
 C20H33NO
Molecular Weight:
 303.48
 Beilstein/REAXYS Number:
 8164441
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More