• USA Home
  • Search Results

  • ferriccitrate26296233805811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ferric citrate

1 Products
Synonym:   Ferric citrate monohydrate, Iron(III) citrate tribasic monohydrate
Image of Ferric citrate
CAS Number:
Empirical Formula (Hill Notation):
 C6H5FeO7 · H2O
Molecular Weight:
 262.96
 Beilstein/REAXYS Number:
 4052503
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More