• USA Home
  • Search Results

  • ferroussulfate27801778263011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ferrous sulfate

2 Products
Synonym:   Ferrous sulfate heptahydrate, Iron(II) sulfate heptahydrate
Image of Ferrous sulfate
CAS Number:
Linear Formula:
 FeSO4 · 7H2O
Molecular Weight:
 278.01
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More