• USA Home
  • Search Results

  • ferulicacid1941853798411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ferulic acid

2 Products
Synonym:   trans-4-Hydroxy-3-methoxycinnamic acid, trans-Ferulic acid, Ferulic acid
Image of Ferulic acid
CAS Number:
Linear Formula:
 HOC6H3(OCH3)CH=CHCO2H
Molecular Weight:
 194.18
 Beilstein/REAXYS Number:
 1570363
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
Show More