• USA Home
  • Search Results

  • fexofenadinehydrochloride5381215343940811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fexofenadine hydrochloride

4 Products
Synonym:   Fexofenidine hydrochloride, MDL 16455 hydrochloride, Terfenidine carboxylate hydrochloride
Image of Fexofenadine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C32H39NO4 · HCl
Molecular Weight:
 538.12
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Anal. Standard Form
  Physical Form
  Special Grade
Show More