• USA Home
  • Search Results

  • fibroblastgrowthfactorfrombovinepituitary123456203154311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fibroblast Growth Factor from bovine pituitary

1 Products
Synonym:   FGF
CAS Number:
 MDL number:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More