• USA Home
  • Search Results

  • flavone2222452582611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Flavone

1 Products
Synonym:   2-Phenyl-4H-1-benzopyran-4-one, 2-Phenylchromone
Image of Flavone
CAS Number:
Empirical Formula (Hill Notation):
 C15H10O2
Molecular Weight:
 222.24
 Beilstein/REAXYS Number:
 157598
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
F2003 Flavone, ≥99.0%,