• USA Home
  • Search Results

  • florisiladsorbentforchromatography10039134388011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Florisil® Adsorbent for Chromatography

2 Products
Synonym:   Activated magnesium silicate, Florisil®, Magnesium silicate
Image of Florisil<SUP>®</SUP> Adsorbent for Chromatography
CAS Number:
Empirical Formula (Hill Notation):
 MgO3Si
Molecular Weight:
 100.39
 EC Number:
 MDL number:
 NACRES:
 SB.52
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Particle Dimension
  Pore size
  Matrix active group
  Physical Form
  Special Grade
Show More