• USA Home
  • Search Results

  • flufenamicacid2812353078911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Flufenamic acid

1 Products
Synonym:   N-(α,α,α-Trifluoro-m-tolyl)anthranilic acid, N-(3-[Trifluoromethyl]phenyl)anthranilic acid
Image of Flufenamic acid
CAS Number:
Linear Formula:
 2-(CF3C6H4NH)C6H4CO2H
Molecular Weight:
 281.23
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More