• USA Home
  • Search Results

  • flumazenil303297875581411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Flumazenil

2 Products
Synonym:   Ro 15-1788, Ro15-1788
Image of Flumazenil
CAS Number:
Empirical Formula (Hill Notation):
 C15H14FN3O3
Molecular Weight:
 303.29
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More