• USA Home
  • Search Results

  • fluocinoloneacetonide452496773211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fluocinolone acetonide

4 Products
Synonym:   6α,9α-Difluoro-11β,16α,17α,21-tetrahydroxy-1,4-pregnadiene-3,20-dione, 6α,9α-Difluoro-16α-hydroxyprednisolone 16,17-acetonide, 6α-Fluorotriamcinolone acetonide, Sinalar, Synandone
Image of Fluocinolone acetonide
CAS Number:
Empirical Formula (Hill Notation):
 C24H30F2O6
Molecular Weight:
 452.49
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Titertest
  Anal. Standard Form
  Physical Form
Show More