• USA Home
  • Search Results

  • fluorescamine278263818312911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fluorescamine

1 Products
Synonym:   4-Phenylspiro-[furan-2(3H),1-phthalan]-3,3′-dione
Image of Fluorescamine
CAS Number:
Empirical Formula (Hill Notation):
 C17H10O4
Molecular Weight:
 278.26
 Beilstein/REAXYS Number:
 921143
 EC Number:
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More