• USA Home
  • Search Results

  • fluoresceinamineisomeri34732332634911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fluoresceinamine, isomer I

1 Products
Synonym:   5-Aminofluorescein
Image of Fluoresceinamine, isomer I
CAS Number:
Empirical Formula (Hill Notation):
 C20H13NO5
Molecular Weight:
 347.32
 Beilstein/REAXYS Number:
 48395
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More