• USA Home
  • Search Results

  • fluoresceinisothiocyanatedextran123456084246811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fluorescein isothiocyanate–dextran

18 Products
Synonym:   FITC–Dextran, FITC-Dextran 4
CAS Number:
 MDL number:
 NACRES
 NA.32
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Extent of labeling
  Assay
  Application(s)
  Fluorescence Test
  Solubility
  Physical Form
  Conjugate
  Biological Source
  Molecular weight
Show More

Product #
Description
46944 Fluorescein isothiocyanate–dextran, average mol wt 4,000, (FITC:Glucose = 1:250),
FD4 Fluorescein isothiocyanate–dextran, average mol wt 3,000-5,000,
46945 Fluorescein isothiocyanate–dextran, average mol wt 70,000, (FITC:Glucose = 1:250),
FD2000S Fluorescein isothiocyanate–dextran, average mol wt 2,000,000,
FD40S Fluorescein isothiocyanate–dextran, average mol wt 40,000,
FD10S Fluorescein isothiocyanate–dextran, average mol wt 10,000,
FD40 Fluorescein isothiocyanate–dextran, average mol wt 40,000,
FD20S Fluorescein isothiocyanate–dextran, average mol wt 20,000,
FD70S Fluorescein isothiocyanate–dextran, average mol wt 60,000-76,000,
FD500S Fluorescein isothiocyanate–dextran, average mol wt 500,000,
FD150S Fluorescein isothiocyanate–dextran, average mol wt 150,000,
46946 Fluorescein isothiocyanate–dextran, average mol wt 150,000, (FITC:Glucose = 1:160),
FD70 Fluorescein isothiocyanate–dextran, average Mw 59,000-77,000,
52471 Fluorescein isothiocyanate–dextran, average mol wt 2,000,000,
FD250S Fluorescein isothiocyanate–dextran, average mol wt 250,000,
FD20 Fluorescein isothiocyanate–dextran, average mol wt 20,000,
90718 Fluorescein isothiocyanate–dextran, average Mw 70,000,
69658 Fluorescein isothiocyanate–dextran, average mol wt 150,000,