• USA Home
  • Search Results

  • fluoresceinisothiocyanateisomeri38938332632711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fluorescein isothiocyanate isomer I

3 Products
Synonym:   FITC, Fluorescein 5-isothiocyanate
Image of Fluorescein isothiocyanate isomer I
CAS Number:
Empirical Formula (Hill Notation):
 C21H11NO5S
Molecular Weight:
 389.38
 Beilstein/REAXYS Number:
 1407295
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Solubility
  Assay
Show More