• USA Home
  • Search Results

  • fluoresceinsodiumsalt3762751847811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fluorescein sodium salt

5 Products
Synonym:   Acid Yellow 73, D&C;Yellow No. 8, NaFl, NaFluo, Sodium fluorescein, Uranine
Image of Fluorescein sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C20H10Na2O5
Molecular Weight:
 376.27
 Beilstein/REAXYS Number:
 3833041
 Colour Index Number:
 45350
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Restriction
  Solubility
  Melting Point
  Special Grade
  pH Value
  Physical Form
Show More