• USA Home
  • Search Results

  • fluorometholone3764642613111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fluorometholone

3 Products
Synonym:   11β,17α-Dihydroxy-9-fluoro-6-methyl-1,4-pregnadiene-3,20-dione
Image of Fluorometholone
CAS Number:
Empirical Formula (Hill Notation):
 C22H29FO4
Molecular Weight:
 376.46
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More