• USA Home
  • Search Results

  • fluoronnnntetramethylformamidiniumhexafluorophosphate2641216429823111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fluoro-N,N,N′,N′-tetramethylformamidinium hexafluorophosphate

1 Products
Synonym:   TFFH
Image of Fluoro-<I>N,N,N′,N′</I>-tetramethylformamidinium hexafluorophosphate
CAS Number:
Linear Formula:
 [FC[=N(CH3)2]N(CH3)2]PF6
Molecular Weight:
 264.12
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More