• USA Home
  • Search Results

  • fluorotrimethylsilane921942056411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fluorotrimethylsilane

1 Products
Synonym:   Trimethylfluorosilane, Trimethylsilyl fluoride
Image of Fluorotrimethylsilane
CAS Number:
Linear Formula:
 (CH3)3SiF
Molecular Weight:
 92.19
 Beilstein/REAXYS Number:
 1731132
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More