• USA Home
  • Search Results

  • fluorouracilimpurityc112096622811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fluorouracil impurity C

1 Products
Synonym:   2,4(1H,3H)-Pyrimidinedione, 2,4-Dihydroxypyrimidine, 2,4-Pyrimidinediol, Uracil
Image of Fluorouracil impurity C
CAS Number:
Empirical Formula (Hill Notation):
 C4H4N2O2
Molecular Weight:
 112.09
 Beilstein/REAXYS Number:
 507828
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More