• USA Home
  • Search Results

  • fluoxetinehydrochloride345795629678711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fluoxetine hydrochloride

6 Products
Synonym:   (±)-N-Methyl-γ-[4-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride, Fluoxetine HCl, LY-110,140 hydrochloride, Prozac®
Image of Fluoxetine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C17H18F3NO · HCl
Molecular Weight:
 345.79
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Solubility
  Manufacturer designation
  Anal. Standard Form
  Physical Form
  Special Grade
Show More