• USA Home
  • Search Results

  • fluvoxamineforsystemsuitability434416171882911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fluvoxamine for system suitability

1 Products
Synonym:   (E)-5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone-O-(2-aminoethyl)oxime maleate, Fluvoxamine maleate
Image of Fluvoxamine for system suitability
CAS Number:
Empirical Formula (Hill Notation):
 C15H21F3N2O2 · C4H4O4
Molecular Weight:
 434.41
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More