• USA Home
  • Search Results

  • fmoc2abzoh3593715025642111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fmoc-2-Abz-OH

1 Products
Synonym:   2-(Fmoc-amino)benzoic acid, N-Fmoc-anthranilic acid
Image of Fmoc-2-Abz-OH
CAS Number:
Empirical Formula (Hill Notation):
 C22H17NO4
Molecular Weight:
 359.37
 Beilstein/REAXYS Number:
 7659726
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
446033 Fmoc-2-Abz-OH, 97%,