• USA Home
  • Search Results

  • fmoccystbuoh399506743613911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Fmoc-Cys(tBu)-OH

2 Products
Synonym:   Fmoc-Cys(tBu)-OH, N-α-Fmoc-S-t.-butyl-L-cysteine
Image of Fmoc-Cys(tBu)-OH
CAS Number:
Empirical Formula (Hill Notation):
 C22H25NO4S
Molecular Weight:
 399.50
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More